Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284411.1 | NP_001271340.1 | 4 | 342 | ||
NM_001284412.1 | NP_001271341.1 | 4 | 318 | |||
NM_001284414.1 | NP_001271343.1 | 4 | 318 | |||
NM_026776.4 | NP_081052.2 | 4 | 318 | |||
GenBank mRNA | AK002500.1 | - | 4 | 280 | ||
AK003715.2 | - | 4 | 290 | |||
AK003877.1 | - | 4 | 276 | |||
AK082933.1 | - | 4 | 297 | |||
AK145811.1 | - | 4 | 289 | |||
AK150822.1 | - | 4 | 300 | |||
AK162230.1 | - | 4 | 291 | |||
AK166008.1 | - | 4 | 297 | |||
AK190530.1 | - | 2 | 72 | |||
AK214794.1 | - | 2 | 72 | |||
BC029181.1 | - | 4 | 282 | 4240500 |