Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145705.3 | NP_663751.2 | 7 | 1303 | ||
GenBank mRNA | AF214013.1 | - | 7 | 1293 | ||
AK011154.1 | - | 7 | 1271 | |||
AK016456.1 | - | 7 | 1237 | |||
AK028952.1 | - | 6 | 1428 | |||
AK090324.1 | - | 7 | 1270 | |||
AK144592.1 | - | 7 | 1232 | |||
AK169480.1 | - | 3 | 1056 | |||
BC030347.1 | - | 7 | 1229 | 5003804 |