Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm70a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138599.5 | NP_613065.2 | 7 | 1350 | ||
GenBank mRNA | AK012084.1 | - | 7 | 1351 | ||
AK083586.1 | - | 7 | 1351 | |||
AK122356.1 | - | 1283 | ||||
AK145458.1 | - | 7 | 1363 | |||
AK150429.1 | - | 7 | 1351 | |||
AK161720.1 | - | 7 | 1340 | |||
AK169809.1 | - | 7 | 1312 | |||
AK217112.1 | - | 154 | ||||
BC057096.1 | - | 7 | 1351 | 5709110 | ||
BC139420.1 | - | 7 | 1323 | 8862442 | ||
BC139421.1 | - | 7 | 1323 | 8862443 |