Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133996.3 | NP_598757.1 | 2 | 695 | ||
GenBank mRNA | AK083289.1 | - | 1 | 831 | ||
BC022137.1 | - | 1 | 762 | |||
BC030734.1 | - | 1 | 635 | |||
BC034832.1 | - | 1 | 794 |