Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013860.2 | NP_038888.2 | 1 | 1713 | ||
GenBank mRNA | AJ132409.1 | - | 1 | 1702 | ||
AK004806.1 | - | 1 | 1017 | |||
AK047198.1 | - | 1 | 1707 | |||
AK083970.1 | - | 1 | 1674 | |||
AK191283.1 | - | 1 | 29 | |||
AK191584.1 | - | 1 | 29 | |||
AK192393.1 | - | 1 | 29 | |||
AK194799.1 | - | 1 | 29 | |||
AK195125.1 | - | 1 | 37 | |||
AK198802.1 | - | 1 | 29 | |||
AK200245.1 | - | 1 | 29 | |||
AK203953.1 | - | 1 | 37 | |||
AK208965.1 | - | 1 | 29 | |||
BC056449.1 | - | 1 | 1588 | 5703026 |