Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013868.4 | NP_038896.2 | 2 | 1258 | ||
GenBank mRNA | AF155909.1 | - | 328 | |||
AJ243192.1 | - | 328 | ||||
AK030025.1 | - | 2 | 1259 | |||
AK086011.1 | - | 2 | 335 | |||
BC046993.1 | - | 2 | 294 | |||
BC089584.1 | - | 2 | 333 | 6755510 |