Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scmh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159630.1 | NP_001153102.1 | 13 | 1910 | ||
NM_013883.2 | NP_038911.1 | 13 | 1910 | |||
GenBank mRNA | AB030906.1 | - | 13 | 1809 | ||
AB221680.1 | - | 9 | 1451 | |||
AB221681.1 | - | 9 | 1451 | |||
AK182691.1 | - | 1 | 168 | |||
AK185818.1 | - | 1 | 167 | |||
BC025478.1 | - | 4 | 2152 | |||
BC034667.1 | - | 11 | 1620 | 5356322 |