Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgfrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013886.4 | NP_038914.2 | 4 | 814 | ||
GenBank mRNA | AB029493.1 | - | 4 | 377 | ||
AK014849.1 | - | 4 | 638 | |||
AK019487.1 | - | 4 | 477 | |||
AK044858.1 | - | 4 | 639 | |||
AK045542.1 | - | 4 | 814 | |||
AK051401.1 | - | 4 | 736 | |||
AK162418.1 | - | 4 | 457 | |||
AK162967.1 | - | 4 | 378 | |||
BC055734.1 | - | 4 | 401 | 6810355 |