Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023141.2 | NP_075630.2 | 4 | 690 | ||
GenBank mRNA | AJ299404.1 | - | 4 | 658 | ||
AK007814.1 | - | 4 | 679 | |||
AK009693.1 | - | 4 | 708 | |||
AK017908.1 | - | 4 | 635 | |||
AK151048.1 | - | 4 | 676 | |||
AK151390.1 | - | 5 | 727 | |||
AK152243.1 | - | 4 | 655 | |||
AK152358.1 | - | 4 | 691 | |||
AK152484.1 | - | 4 | 691 | |||
AK153391.1 | - | 4 | 676 | |||
AK170296.1 | - | 4 | 685 | |||
AK172355.1 | - | 4 | 665 | |||
BC025525.1 | - | 4 | 674 | |||
BC052851.1 | - | 4 | 979 | |||
BC071216.1 | - | 4 | 671 | 30439199 |