Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013923.2 | NP_038951.1 | 9 | 2119 | ||
GenBank mRNA | AF120206.1 | - | 2012 | |||
AF120207.1 | - | 8 | 2012 | |||
AK147768.1 | - | 9 | 2107 | |||
BC040769.1 | - | 8 | 1894 | 4457807 | ||
DV656092.1 | - | 4 | 603 | |||
X71642.1 | - | 5 | 666 |