Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098511.2 | NP_001091981.1 | 19 | 2251 | ||
NM_001243952.1 | NP_001230881.1 | 19 | 2340 | |||
NM_001243953.1 | NP_001230882.1 | 18 | 2080 | |||
NM_004520.4 | NP_004511.2 | 18 | 2137 | |||
GenBank mRNA | AK297347.1 | - | 19 | 1997 | ||
AK302270.1 | - | 18 | 1774 | |||
AK315866.1 | - | 18 | 1764 | |||
AY317140.1 | - | 16 | 1668 | |||
BC031828.2 | - | 17 | 1762 | 4932771 | ||
EF560716.1 | - | 19 | 1962 | |||
EF560728.1 | - | 18 | 1842 | |||
Y08319.1 | - | 17 | 1763 |