Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Postn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198765.1 | NP_001185694.1 | 18 | 2145 | ||
NM_001198766.1 | NP_001185695.1 | 17 | 2064 | |||
NM_015784.3 | NP_056599.1 | 17 | 2064 | |||
GenBank mRNA | AK030756.1 | - | 18 | 2144 | ||
AK154049.1 | - | 17 | 2062 | |||
AK163070.1 | - | 1 | 4053 | |||
AY651928.1 | - | 18 | 2115 | |||
BC007141.1 | - | 18 | 2139 | |||
BC031449.1 | - | 17 | 2033 | 4457222 | ||
D13664.1 | - | 17 | 2046 | |||
E05655.1 | - | 17 | 2060 | |||
KM117171.1 | - | 17 | 2028 | |||
KM117172.1 | - | 18 | 2109 |