Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIFC3 SIRNA Assays ›
Gene Symbol:
KIFC3
Gene Name:
kinesin family member C3
Gene Aliases:
-
Chromosome Location:
Chr.16: 57758217 - 57863053 on Build GRCh38
UniGene ID:
Hs.23131
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s7844
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIFC3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130099.1
NP_001123571.1
7
1214
NM_001130100.1
NP_001123572.1
8
1270
NM_001318710.1
NP_001305639.1
8
1151
NM_001318711.1
NP_001305640.1
8
1312
NM_001318712.1
NP_001305641.1
6
887
NM_001318713.1
NP_001305642.1
6
1289
NM_001318714.1
NP_001305643.1
9
1659
NM_001318715.1
NP_001305644.1
8
1248
NM_005550.3
NP_005541.3
8
1270
NR_134678.1
8
1454
XM_005255937.1
XP_005255994.1
10
1779
XM_005255944.1
XP_005256001.1
8
1133
XM_006721188.1
XP_006721251.1
11
1857
XM_011523075.1
XP_011521377.1
8
1437
XM_011523076.1
XP_011521378.1
8
1437
XM_011523077.1
XP_011521379.1
8
1236
XM_011523078.1
XP_011521380.1
8
1039
XM_011523079.1
XP_011521381.1
8
1374
XM_017023219.1
XP_016878708.1
11
1826
XM_017023220.1
XP_016878709.1
11
2003
XM_017023221.1
XP_016878710.1
10
1779
XM_017023222.1
XP_016878711.1
12
2021
XM_017023223.1
XP_016878712.1
8
1154
XM_017023224.1
XP_016878713.1
10
1688
XM_017023225.1
XP_016878714.1
6
2258
XM_017023226.1
XP_016878715.1
9
1531
XM_017023227.1
XP_016878716.1
9
1531
GenBank mRNA
AB209290.1
-
8
1205
AF004426.1
-
4
656
AK091891.1
-
407
AK291737.1
-
7
1109
AK295926.1
-
3
232
AK296995.1
-
8
1150
AK299431.1
-
8
1212
AK302743.1
-
6
887
AK303021.1
-
6
1217
AK304132.1
-
8
1258
AK307846.1
-
8
1228
BC001211.1
-
8
1239
3503847
BC008014.1
-
2
235
BC034234.1
-
9
1599
5296397
BC041132.1
-
8
1100
5760988
BC047051.1
-
8
1398
5297265
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00194304_m1
Panther Classification:
Molecular Function -
microtubule binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule-based movement
Golgi organization
visual perception
zonula adherens maintenance
epithelial cell-cell adhesion
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, minus-end-directed
ATPase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0