Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myadm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093764.1 | NP_001087233.1 | 3 | 1151 | ||
NM_001093765.1 | NP_001087234.1 | 3 | 1100 | |||
NM_001093766.1 | NP_001087235.1 | 2 | 1038 | |||
NM_016969.2 | NP_058665.2 | 3 | 939 | |||
GenBank mRNA | AJ001616.1 | - | 3 | 959 | ||
AK052258.1 | - | 3 | 1058 | |||
AK053414.1 | - | 3 | 891 | |||
AK089538.1 | - | 3 | 922 | |||
AK149962.1 | - | 3 | 1082 | |||
AK159160.1 | - | 1082 | ||||
BC056355.1 | - | 3 | 1034 | 6811773 | ||
BC119364.1 | - | 2 | 822 | 8734113 | ||
BC119368.1 | - | 2 | 819 | 8734117 |