Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081475.1 | NP_001074944.1 | 3, 4 | 320 | ||
NM_016777.3 | NP_058057.3 | 4, 5 | 314 | |||
GenBank mRNA | AF034610.2 | - | 4, 5 | 304 | ||
AK083333.1 | - | 3, 4 | 321 | |||
AK132690.1 | - | 4, 5 | 293 | |||
AK161367.1 | - | 4, 5 | 293 | |||
BC004693.1 | - | 3, 4 | 287 | 3498591 |