Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016856.3 | NP_058552.1 | 8 | 1296 | ||
GenBank mRNA | AF012822.1 | - | 8 | 1282 | ||
AK140869.1 | - | 8 | 1278 | |||
AK141325.1 | - | 8 | 1278 | |||
AK147746.1 | - | 8 | 1297 | |||
BC007163.1 | - | 6 | 954 | |||
BC013628.1 | - | 8 | 1271 | 3497854 |