Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002231.2 | NP_001002231.1 | 5 | 1686 | ||
NM_001256080.1 | NP_001243009.1 | 4 | 1489 | |||
NM_005551.4 | NP_005542.1 | 5 | 1649 | |||
NR_045762.1 | 5 | 1673 | ||||
NR_045763.1 | 5 | 1735 | ||||
GenBank mRNA | AF188745.1 | - | 5 | 1648 | ||
AF188746.1 | - | 5 | 1624 | |||
AF188747.1 | - | 5 | 1661 | |||
BX641830.1 | - | 1 | 273 |