Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009818.1 | NP_001009818.1 | 2 | 159 | ||
GenBank mRNA | AK028475.1 | - | 2 | 91 | ||
AK166517.1 | - | 2 | 180 | |||
AK171368.1 | - | 2 | 180 | |||
BC031456.1 | - | 2 | 66 | 4037515 | ||
BC062206.1 | - | 2 | 200 | 5150858 | ||
BC064466.1 | - | 2 | 159 | 6825549 |