Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177471.4 | NP_803422.2 | 5 | 418 | ||
GenBank mRNA | AK008760.1 | - | 5 | 389 | ||
AK079810.1 | - | 5 | 419 | |||
AK135942.1 | - | 5 | 395 | |||
AK170686.1 | - | 5 | 368 | |||
BC096763.1 | - | 5 | 376 | 30053993 |