Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145940.2 | NP_666052.1 | 4 | 493 | ||
GenBank mRNA | AK031672.1 | - | 4 | 502 | ||
AK034907.1 | - | 4 | 537 | |||
AK045131.1 | - | 4 | 502 | |||
AK050495.1 | - | 4 | 501 | |||
AK159608.1 | - | 4 | 522 | |||
AK170563.1 | - | 4 | 537 | |||
AK186984.1 | - | 2 | 165 | |||
AK203625.1 | - | 2 | 165 | |||
BC024811.1 | - | 4 | 461 | |||
BC024883.1 | - | 4 | 457 | |||
BC025560.1 | - | 4 | 493 | 5322999 |