Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114396.1 | NP_001107868.1 | 7 | 651 | ||
NM_002262.3 | NP_002253.1 | 6 | 714 | |||
NM_007334.2 | NP_031360.1 | 5 | 621 | |||
XM_006719067.3 | XP_006719130.1 | 6 | 706 | |||
XM_011520650.2 | XP_011518952.1 | 7 | 648 | |||
XM_011520651.2 | XP_011518953.1 | 6 | 705 | |||
XM_017019285.1 | XP_016874774.1 | 6 | 709 | |||
XM_017019286.1 | XP_016874775.1 | 7 | 639 | |||
XM_017019287.1 | XP_016874776.1 | 7 | 656 | |||
XM_017019288.1 | XP_016874777.1 | 5 | 627 | |||
XM_017019289.1 | XP_016874778.1 | 5 | 618 | |||
XM_017019290.1 | XP_016874779.1 | 8 | 2515 | |||
XM_017019291.1 | XP_016874780.1 | 7 | 638 | |||
GenBank mRNA | AB009597.1 | - | 5 | 398 | ||
AF498040.1 | - | 6 | 532 | |||
AF498041.1 | - | 5 | 433 | |||
AJ000001.1 | - | 6 | 485 | |||
AK291506.1 | - | 7 | 656 | |||
AY227806.1 | - | 7 | 651 | |||
AY327501.1 | - | 5 | 519 | |||
BC028009.1 | - | 6 | 527 | 5213081 | ||
U30610.1 | - | 6 | 538 |