Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zwint | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293683.1 | NP_001280612.1 | 4 | 411 | ||
NM_001293684.1 | NP_001280613.1 | 4 | 411 | |||
NM_025635.4 | NP_079911.1 | 4 | 411 | |||
GenBank mRNA | AK004305.1 | - | 4 | 363 | ||
AK008144.1 | - | 4 | 361 | |||
AK013556.1 | - | 4 | 346 | |||
AK075977.1 | - | 4 | 357 | |||
AK082215.1 | - | 4 | 360 | |||
AK134380.1 | - | 4 | 363 | |||
AK182584.1 | - | 2 | 182 | |||
BC013559.1 | - | 4 | 329 | |||
BC027100.1 | - | 4 | 368 | 5359154 | ||
BC034870.1 | - | 4 | 340 | 1382192 |