Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289776.1 | NP_001276705.1 | 1 | 709 | ||
NM_001289777.1 | NP_001276706.1 | 2 | 534 | |||
NM_027530.3 | NP_081806.1 | 1 | 709 | |||
GenBank mRNA | AK013185.1 | - | 2 | 535 | ||
AK018184.1 | - | 1 | 687 | |||
AK045610.1 | - | 1 | 643 | |||
AK083242.1 | - | 1 | 644 | |||
BC017648.1 | - | 1 | 98 | 4527858 | ||
BC049127.1 | - | 1 | 648 | 6821567 | ||
BC058259.1 | - | 1 | 428 | 6469137 | ||
CT010404.1 | - | 1 | 82 | |||
DV068924.1 | - | 2 | 465 |