Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004523.3 | NP_004514.2 | 20 | 3119 | ||
GenBank mRNA | AB208964.1 | - | 6 | 854 | ||
AK314258.1 | - | 20 | 3022 | |||
BC050421.1 | - | 2 | 336 | |||
BC126211.1 | - | 20 | 2918 | 8991927 | ||
BC136474.1 | - | 20 | 2918 | 9020463 | ||
U37426.1 | - | 20 | 2901 | |||
X85137.1 | - | 20 | 2824 |