Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF534114.1 | - | 2, 3 | 287 | ||
AK018166.1 | - | 3, 4 | 338 | |||
AK086301.1 | - | 3, 4 | 497 | |||
AK134581.1 | - | 3, 4 | 341 | |||
AK153400.1 | - | 3, 4 | 325 | |||
BC005640.1 | - | 3, 4 | 326 | |||
BC018178.1 | - | 3, 4 | 312 | |||
BC037451.1 | - | 2, 3 | 316 | 4018648 | ||
U27462.1 | - | 3, 4 | 534 |