Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF534114.1 | - | 1 | 76 | ||
AK086301.1 | - | 2 | 286 | |||
AK134581.1 | - | 2 | 130 | |||
AK153400.1 | - | 2 | 114 | |||
BC005640.1 | - | 2 | 115 | |||
BC018178.1 | - | 2 | 101 | |||
BC037451.1 | - | 1 | 105 | 4018648 | ||
U27462.1 | - | 2 | 323 |