Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016784.3 | NP_058064.2 | 6, 7 | 598 | ||
GenBank mRNA | AF044334.1 | - | 5, 6 | 490 | ||
AK075993.1 | - | 6, 7 | 586 | |||
AK076007.1 | - | 6, 7 | 576 | |||
AK077197.1 | - | 6, 7 | 599 | |||
AK080068.1 | - | 6, 7 | 605 | |||
BC006750.1 | - | 6, 7 | 568 | 3709134 | ||
BC052895.1 | - | 6, 7 | 556 |