Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016786.3 | NP_058066.2 | 3, 4 | 447 | ||
GenBank mRNA | AB011081.1 | - | 3, 4 | 385 | ||
AK018366.1 | - | 3, 4 | 421 | |||
AK035371.1 | - | 3, 4 | 344 | |||
AK036068.1 | - | 3, 4 | 388 | |||
AK048194.1 | - | 3, 4 | 438 | |||
AK139564.1 | - | 3, 4 | 464 | |||
AK160728.1 | - | 3, 4 | 407 | |||
AK162142.1 | - | 3, 4 | 388 | |||
AK166647.1 | - | 3, 4 | 356 | |||
BC002013.1 | - | 3, 4 | 295 | 3496393 | ||
BC085311.1 | - | 3, 4 | 373 | 30148009 |