Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199023.1 | NP_001185952.1 | 13 | 2623 | ||
NM_001199024.1 | NP_001185953.1 | 12 | 2458 | |||
NM_001199025.1 | NP_001185954.1 | 13 | 1963 | |||
NM_016915.4 | NP_058611.1 | 12 | 2004 | |||
GenBank mRNA | AF259401.1 | - | 12 | 1903 | ||
AK144495.1 | - | 13 | 1837 | |||
AK153333.1 | - | 12 | 2264 | |||
BC003487.1 | - | 12 | 1797 | 2811357 | ||
BC049778.1 | - | 12 | 2184 | |||
BC052845.1 | - | 12 | 1829 | 30054573 | ||
BC057209.1 | - | 13 | 2216 | 5329147 | ||
U88624.2 | - | 11 | 1680 |