Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bysl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016859.3 | NP_058555.3 | 4 | 954 | ||
GenBank mRNA | AF007802.1 | - | 2 | 183 | ||
AK019239.1 | - | 4 | 689 | |||
AK032993.1 | - | 4 | 686 | |||
AK049195.1 | - | 4 | 947 | |||
AK051704.1 | - | 4 | 944 | |||
AK083033.1 | - | 4 | 941 | |||
AK143027.1 | - | 4 | 955 | |||
AK168301.1 | - | 4 | 692 | |||
BC017530.1 | - | 4 | 681 | 4925307 |