Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htatip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146049.1 | NP_001139521.1 | 6 | 927 | ||
NM_001146050.1 | NP_001139522.1 | 6 | 761 | |||
NM_001146052.1 | NP_001139524.1 | 5 | 751 | |||
NM_001146053.1 | NP_001139525.1 | 6 | 705 | |||
NM_016865.3 | NP_058561.2 | 6 | 871 | |||
GenBank mRNA | AF061972.1 | - | 5 | 855 | ||
AK008630.1 | - | 5 | 746 | |||
AK144837.1 | - | 6 | 905 | |||
AK146400.1 | - | 5 | 566 | |||
AK151254.1 | - | 6 | 792 | |||
AK152418.1 | - | 6 | 792 | |||
BC004083.1 | - | 6 | 824 | 3593077 | ||
BC017372.1 | - | 5 | 717 | 4013057 |