Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec61a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016906.4 | NP_058602.1 | 7 | 655 | ||
GenBank mRNA | AB032902.1 | - | 7 | 667 | ||
AF145253.1 | - | 7 | 577 | |||
AF222743.1 | - | 7 | 577 | |||
AK054318.1 | - | 2 | 1491 | |||
AK088473.1 | - | 7 | 656 | |||
AK153055.1 | - | 7 | 645 | |||
AK159349.1 | - | 7 | 643 | |||
AK167484.1 | - | 7 | 585 | |||
AK197110.1 | - | 3 | 267 | |||
BC003707.1 | - | 7 | 581 | 3592772 | ||
BC006020.1 | - | 3 | 240 |