Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002267.3 | NP_002258.2 | 15 | 1766 | ||
XM_017020561.1 | XP_016876050.1 | 15 | 1449 | |||
GenBank mRNA | AF005263.1 | - | 15 | 1363 | ||
AF034756.1 | - | 15 | 1353 | |||
AK223480.1 | - | 15 | 1388 | |||
AK290528.1 | - | 15 | 1426 | |||
AK291000.1 | - | 15 | 1390 | |||
BC017355.2 | - | 15 | 1375 | 5020312 | ||
BC024202.2 | - | 15 | 1375 | 3531069 | ||
BC035090.2 | - | 15 | 1767 | 5261465 | ||
D89618.1 | - | 15 | 1442 | |||
Y12394.1 | - | 15 | 1366 |