Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252614.1 | NP_001239543.1 | 5 | 743 | ||
NM_016893.5 | NP_058589.2 | 4 | 654 | |||
GenBank mRNA | AB025198.1 | - | 1 | 343 | ||
AK048520.1 | - | 4 | 792 | |||
AK051811.1 | - | 4 | 611 | |||
AK134537.1 | - | 4 | 747 | |||
AK163494.1 | - | 4 | 770 | |||
BC010666.1 | - | 5 | 722 | 3594582 | ||
BU702757.1 | - | 3 | 592 |