Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113486.1 | NP_001106958.1 | 12 | 1725 | ||
NM_001113487.1 | NP_001106959.1 | 11 | 1905 | |||
NM_001113488.1 | NP_001106960.1 | 10 | 1074 | |||
NM_017380.2 | NP_059076.1 | 10 | 1055 | |||
GenBank mRNA | AJ250723.1 | - | 10 | 1061 | ||
AK031757.1 | - | 10 | 950 | |||
AK122415.2 | - | 11 | 1699 | |||
AK141312.1 | - | 12 | 1666 | |||
AK164581.1 | - | 9 | 3669 | |||
AK210979.1 | - | 2 | 29 | |||
BC046524.1 | - | 12 | 1725 | 5290621 | ||
BC094455.1 | - | 13 | 1429 |