Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017393.2 | NP_059089.1 | 6 | 728 | ||
GenBank mRNA | AJ005253.1 | - | 6 | 696 | ||
AK002238.1 | - | 6 | 667 | |||
AK004024.1 | - | 6 | 677 | |||
AK145765.1 | - | 6 | 690 | |||
AK147719.1 | - | 6 | 656 | |||
AK168053.1 | - | 6 | 675 | |||
AK179220.1 | - | 3 | 297 | |||
BC001998.1 | - | 6 | 687 | 3487489 |