Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002271.4 | NP_002262.3 | 17 | 1921 | ||
XM_005254052.3 | XP_005254109.1 | 16 | 1831 | |||
XM_005254053.3 | XP_005254110.1 | 16 | 1848 | |||
XM_011521087.2 | XP_011519389.1 | 18 | 2012 | |||
XM_011521088.2 | XP_011519390.1 | 18 | 2078 | |||
XM_011521089.2 | XP_011519391.1 | 19 | 2135 | |||
XM_011521090.2 | XP_011519392.1 | 17 | 1971 | |||
XM_017020562.1 | XP_016876051.1 | 17 | 1952 | |||
GenBank mRNA | AK125031.1 | - | 4 | 1400 | ||
AK225286.1 | - | 14 | 1706 | |||
AK302812.1 | - | 14 | 1693 | |||
AK303494.1 | - | 12 | 1334 | |||
BC001497.2 | - | 14 | 1689 | 2823581 | ||
BC019309.1 | - | 14 | 1689 | |||
BC045640.1 | - | 17 | 1902 | 5298410 | ||
BX648222.1 | - | 14 | 1751 | |||
U72761.1 | - | 14 | 1687 | |||
Y08890.1 | - | 17 | 1923 |