Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uevld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040695.1 | NP_001035785.1 | 7 | 762 | ||
GenBank mRNA | AK154536.1 | - | 7 | 713 | ||
AK155685.1 | - | 7 | 709 | |||
BC054796.1 | - | 7 | 768 | 6843964 | ||
CB524764.1 | - | 7 | 768 | |||
CN538811.1 | - | 6 | 632 |