Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113517.1 | NP_001106989.1 | 14 | 1721 | ||
NM_001113518.1 | NP_001106990.1 | 13 | 1510 | |||
NM_017402.4 | NP_059098.2 | 13 | 1510 | |||
GenBank mRNA | AF247654.1 | - | 13 | 1406 | ||
AF247655.1 | - | 11 | 1127 | |||
AF343877.1 | - | 13 | 1390 | |||
AK052545.1 | - | 13 | 1481 | |||
AK129064.1 | - | 14 | 1657 | |||
AK139156.1 | - | 14 | 1650 | |||
AK164308.1 | - | 1 | 2649 | |||
AY220301.1 | - | 13 | 1475 | |||
BC044838.1 | - | 13 | 1495 | 3499604 | ||
U96634.1 | - | 11 | 1200 |