Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113518.1 | NP_001106990.1 | 20 | 2370 | ||
NM_017402.4 | NP_059098.2 | 19 | 2193 | |||
GenBank mRNA | AF247654.1 | - | 20 | 2266 | ||
AF247655.1 | - | 17 | 1762 | |||
AK047259.1 | - | 2 | 1379 | |||
AK052545.1 | - | 19 | 2164 | |||
AY220301.1 | - | 20 | 2335 | |||
BC044838.1 | - | 19 | 2178 | 3499604 | ||
U96634.1 | - | 17 | 1883 |