Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001150749.1 | NP_001144221.1 | 4 | 953 | ||
NM_017473.4 | NP_059501.1 | 4 | 925 | |||
GenBank mRNA | AB015165.1 | - | 4 | 1041 | ||
AB032058.1 | - | 4 | 833 | |||
AF056194.1 | - | 4 | 930 | |||
BC024603.1 | - | 4 | 903 | 4196075 | ||
BC092255.1 | - | 4 | 879 | 5100891 | ||
BC093514.1 | - | 4 | 867 | 5132290 | ||
CB951082.1 | - | 1 | 52 |