Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017480.2 | NP_059508.2 | 3 | 519 | ||
GenBank mRNA | AB023132.1 | - | 3 | 503 | ||
AF216748.1 | - | 3 | 474 | |||
AF257230.1 | - | 3 | 504 | |||
AJ250559.1 | - | 3 | 519 | |||
AK030827.1 | - | 3 | 512 | |||
BC034852.1 | - | 3 | 506 | 1364228 |