Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291141.1 | NP_001278070.1 | 8 | 1349 | ||
NM_001291142.1 | NP_001278071.1 | 8 | 1358 | |||
NM_001291143.1 | NP_001278072.1 | 10 | 1549 | |||
NM_019400.3 | NP_062273.2 | 9 | 1478 | |||
GenBank mRNA | AB001750.1 | - | 10 | 1385 | ||
AF248489.1 | - | 8 | 1144 | |||
AF248490.1 | - | 8 | 1135 | |||
AK134587.1 | - | 9 | 1479 | |||
AK137236.1 | - | 9 | 1443 | |||
BC003921.1 | - | 1 | 204 | |||
BC060166.1 | - | 9 | 1496 | |||
BC080274.1 | - | 9 | 1450 | |||
BC129853.1 | - | 9 | 1297 | 40109688 | ||
BC129854.1 | - | 9 | 1297 | 40109689 | ||
D86066.1 | - | 9 | 1314 |