Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291141.1 | NP_001278070.1 | 6 | 1024 | ||
NM_001291142.1 | NP_001278071.1 | 6 | 1033 | |||
NM_001291143.1 | NP_001278072.1 | 8 | 1224 | |||
NM_019400.3 | NP_062273.2 | 7 | 1153 | |||
GenBank mRNA | AB001750.1 | - | 8 | 1060 | ||
AF248489.1 | - | 6 | 819 | |||
AF248490.1 | - | 6 | 810 | |||
AK134587.1 | - | 7 | 1153 | |||
AK137236.1 | - | 7 | 1118 | |||
BC009814.1 | - | 7 | 1149 | |||
BC060166.1 | - | 7 | 1132 | |||
BC080274.1 | - | 7 | 1126 | |||
BC129853.1 | - | 7 | 972 | 40109688 | ||
BC129854.1 | - | 7 | 972 | 40109689 | ||
D86066.1 | - | 7 | 989 |