Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017476.2 | NP_059504.2 | 2, 3 | 158 | ||
GenBank mRNA | AB028921.1 | - | 2, 3 | 151 | ||
AK011325.1 | - | 2, 3 | 163 | |||
AK166526.1 | - | 2, 3 | 176 | |||
AK203044.1 | - | 1, 2 | 31 | |||
AK206808.1 | - | 1, 2 | 31 | |||
BC010401.1 | - | 2, 3 | 208 | |||
BC046801.1 | - | 2, 3 | 147 | 6403966 | ||
BC051015.1 | - | 2, 3 | 158 |