Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017465.2 | NP_059493.2 | 4 | 567 | ||
GenBank mRNA | AF026072.1 | - | 4 | 534 | ||
AF478566.1 | - | 3 | 690 | |||
BC009811.1 | - | 4 | 609 | 4008307 | ||
BC009813.1 | - | 4 | 497 | 4039050 |