Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacng6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133183.1 | NP_573446.1 | 1 | 198 | ||
GenBank mRNA | AF361348.1 | - | 1 | 198 | ||
BC115456.1 | - | 1 | 198 | 40100157 | ||
BC115457.1 | - | 1 | 198 | 40100159 |