Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195083.1 | NP_001182012.1 | 5 | 774 | ||
NM_001195130.1 | NP_001182059.1 | 13 | 2174 | |||
NM_018774.4 | NP_061244.1 | 13 | 2167 | |||
GenBank mRNA | AB062362.1 | - | 5 | 760 | ||
AF060076.1 | - | 6 | 753 | |||
BC051657.1 | - | 5 | 667 | |||
BC057571.1 | - | 13 | 2110 | 6410302 | ||
BC071246.1 | - | 5 | 755 | |||
U81491.1 | - | 12 | 2064 |