Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195083.1 | NP_001182012.1 | 2 | 463 | ||
NM_001195130.1 | NP_001182059.1 | 10 | 1863 | |||
NM_018774.4 | NP_061244.1 | 10 | 1856 | |||
GenBank mRNA | AB062362.1 | - | 2 | 449 | ||
AF060076.1 | - | 3 | 442 | |||
BC057571.1 | - | 10 | 1799 | 6410302 | ||
BC071246.1 | - | 2 | 444 | |||
U81491.1 | - | 9 | 1753 |