Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019443.2 | NP_062316.1 | 2 | 306 | ||
GenBank mRNA | AK007840.1 | - | 2 | 213 | ||
AK131584.1 | - | 505 | ||||
BC024364.1 | - | 2 | 215 | 5031206 | ||
CF383819.1 | - | 1 | 90 | |||
CT010398.1 | - | 2 | 158 | |||
Y07708.1 | - | 2 | 325 |